Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141299

Catalog No. 01-675-640


Add to cart

Description

Description

Suggested positive control: antigen standard for TNFRSF21 (transient overexpression lysate), Daudi, HeLa, or Jurkat.

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel death domain containing receptor was recently identified and designated DR6 (for death receptor 6) (1). The extracellular domain of DR6 contains four TNFR-like cysteine rich motifs that are related to those of osteoprotegerin (OPG) (2) and TNFR2 (3) with 36% and 42% amino acid identities, respectively. The death domain of DR6 (amino acids 428-494) located adjacent to the transmembrane domain is most related to TNFR1 (27.2%) and least to DR5 (19.7%) (1).
Specifications

Specifications

DR6
Polyclonal
Unconjugated
TNFRSF21
Liquid
TNFRSF21
Synthetic peptide corresponding to residues 371-655 of human DR6.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
O75509
BM-018, DR6, MGC31965
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
27242
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit