Learn More
anti-DR6/TNFRSF21, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NB6001400SS
 View more versions of this product
Catalog No. NB6001400SS
Add to cart
Description
Description
DR6/TNFRSF21 Polyclonal antibody specifically detects DR6/TNFRSF21 in Human samples. It is validated for Western Blot.
Specifications
Specifications
DR6/TNFRSF21
0.5 mg/mL
Western Blot 2 - 4 ug/ml
NP_043000
BM-018, CD358 antigen, Death receptor 6, DR6TNFR-related death receptor 6, MGC31965, tumor necrosis factor receptor superfamily member 21, tumor necrosis factor receptor superfamily, member 21
Rabbit
IgG
0.025 mg
Apoptosis, Cell Biology
Primary
27242
Western Blot
Unconjugated
PBS containing 0.05% BSA with 0.05% Sodium Azide
Purified
TNFRSF21
The antibody was raised against the cytoplasmic domain (amino acids 371-655) of human DR6 protein.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human