Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRAK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527694
Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Zebrafish (80%), Xenopus laevis (84%), Rabbit (95%), Chicken (89%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DRAK1 Polyclonal antibody specifically detects DRAK1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
DRAK1 | |
Polyclonal | |
Unconjugated | |
STK17A | |
DAP kinase-related apoptosis-inducing protein kinase 1; death-associated protein kinase-related 1; DRAK1; serine/threonine kinase 17a; serine/threonine kinase 17a (apoptosis-inducing); serine/threonine-protein kinase 17A; STK17A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9263 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.91 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9UEE5 | |
STK17A | |
Recombinant fragment corresponding to a region within amino acids 47 and 230 of Human DRAK1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction