Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DRAK2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP151948

Catalog No. NBP151948

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

DRAK2
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
STK17B
Peptide with sequence C-HSVRSSEDKTSK corresponding to internal region according to NP_004217.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody may cross-react with rat STK17B (GeneID: 170904).
ELISA, Western Blot
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
NP_004217.1
DAP kinase-related apoptosis-inducing protein kinase 2, DRAK2death-associated protein kinase-related 2, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 17b, serine/threonine kinase 17b (apoptosis-inducing), serine/threonine-protein kinase 17B
Goat
IgG
0.1 mg
Apoptosis
Primary
9262
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only