Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DRAK2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $293.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This polyclonal antibody was raised against a peptide corresponding to amino acids 351 to 365 of human DRAK2 protein.
Antigen DRAK2
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056695T
View Documents
Novus Biologicals
NB10056695SS
0.025 mg
Each for $119.60
Add to cart
 
NB10056695
View Documents
Novus Biologicals
NB10056695
0.1 mg
Each for $293.00
Add to cart
 
Description

Description

DRAK2 Polyclonal antibody specifically detects DRAK2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Rabbit
This polyclonal antibody was raised against a peptide corresponding to amino acids 351 to 365 of human DRAK2 protein.
Polyclonal
Primary
Western Blot
9262
IgG
DRAK2
Purified
RUO
NP_004217.1
PBS with 0.02% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit