Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRAM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520336
Description
A suggested positive control is K562 cell lysate. PA5-20336 can be used with blocking peptide PEP-0456.
DRAM Polyclonal antibody specifically detects DRAM in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
DRAM | |
Polyclonal | |
Unconjugated | |
DRAM1 | |
1200002N14Rik; Damage-regulated autophagy modulator; DNA damage regulated autophagy modulator 1; DNA damage-regulated autophagy modulator protein 1; DNA-damage regulated autophagy modulator 1; DRAM; Dram1; FLJ11259; hypothetical protein FLJ11259; zgc:101811 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55332, 679937, 71712 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8N682, Q9DC58 | |
DRAM1 | |
A 16 amino acid peptide from near the amino terminus of human DRAM. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction