Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRBP76 Mouse anti-Human, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612154

 View more versions of this product

Catalog No. BDB612154


Add to cart

Description

Description

The double stranded RNA (dsRNA)-dependent, Ser/Thr protein kinase, PKR, is encoded by an IFN-inducible gene and is critical for the anti-viral responses mediated by IFN. Interaction with activators such as heparin, dsRNA, and the dsRNA-binding proteins (DRBPs), DRBP76, PACT, and RAX, induces PKR autophosphorylation and activation. DRBP76 was identified through its binding to dsRNA and PKR. In addition, DRBP76 has been identified as the alternatively spliced nuclear phosphoproteins of 90kDa (NFAR-1/NF90) and 110kDa (NFAR-2), as well as M-phase phosphoprotein (MPP4), translational control protein 80 (TCP80), and interleukin enhancer binding factor 3 (ILF3). DRBP76 contains a bi-partite nuclear localization signal at amino acids 369-373 and 386-394, two DRB domains in the C-terminal half, and a C-terminal RG2 domain that is present in many RNA binding proteins. DRBP76 binds pre-mRNAs and spliced mRNAs, co-localizes with PKR in the nucleus, and is phosphorylated by PKR and possibly cyclin-dependent kinases. Thus, DRBP76 may be one of multiple DRBP variants, which associate with PKR and regulate RNA translation.

Immunofluorescence, Western Blotting

Specifications

Specifications

DRBP76
21
Unconjugated
Affinity Purified
Human DRBP76 aa. 592-695
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.