Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRBP76 Mouse anti-Human, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

$463.00

Specifications

Antigen DRBP76
Isotype IgG1
Immunogen Human DRBP76 aa. 592-695
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB612155
View Documents Promotion Details
BD Biosciences
612155
150μg
Each for $463.00
Add to cart
 
BDB612154
View Documents Promotion Details
BD Biosciences
612154
50μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

The double stranded RNA (dsRNA)-dependent, Ser/Thr protein kinase, PKR, is encoded by an IFN-inducible gene and is critical for the anti-viral responses mediated by IFN. Interaction with activators such as heparin, dsRNA, and the dsRNA-binding proteins (DRBPs), DRBP76, PACT, and RAX, induces PKR autophosphorylation and activation. DRBP76 was identified through its binding to dsRNA and PKR. In addition, DRBP76 has been identified as the alternatively spliced nuclear phosphoproteins of 90kDa (NFAR-1/NF90) and 110kDa (NFAR-2), as well as M-phase phosphoprotein (MPP4), translational control protein 80 (TCP80), and interleukin enhancer binding factor 3 (ILF3). DRBP76 contains a bi-partite nuclear localization signal at amino acids 369-373 and 386-394, two DRB domains in the C-terminal half, and a C-terminal RG2 domain that is present in many RNA binding proteins. DRBP76 binds pre-mRNAs and spliced mRNAs, co-localizes with PKR in the nucleus, and is phosphorylated by PKR and possibly cyclin-dependent kinases. Thus, DRBP76 may be one of multiple DRBP variants, which associate with PKR and regulate RNA translation.

Immunofluorescence, Western Blotting

Specifications

Specifications

DRBP76
Human DRBP76 aa. 592-695
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
21
250μg/mL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit