Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dsk2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512077

Catalog No. PIPA512077

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Dsk2 increases the half-life of proteins destined to be degraded by the proteasome, and may modulate proteasome-mediated protein degradation. The Dsk2 protein binds UBE3A and BTRC, and interacts with the 19S proteasome subunit. In the cytoplasm, Dsk2 colocalizes with the proteasome; it is also associated with fibers in mitotic cells in the nucleus. Dsk2 is highly expressed in mitotic cells from metaphase to telophase, while expression in non-mitotic cells is very low.
Specifications

Specifications

Dsk2
Polyclonal
PBS with 0.09% sodium azide
Q9UHD9
Ubiquilin 2; Protein linking IAP with cytoskeleton-2; PLIC-2; hPLIC-2; Ubiquitin-like product Chap1/Dsk2; DSK2 homolog; Chap1; HRIHFB2157
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29978
Western Blot
Unconjugated
UBQLN2
Liquid
UBQLN2
KLH conjugated synthetic peptide between 515-545 amino acids from the C-terminal region of human Dsk2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit