Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Destrin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526197

Catalog No. PIPA526197

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, chicken, mouse, non-human primate, rat and rabbit based on sequence homology.

DSTN belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This protein encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin).
Specifications

Specifications

Destrin
Polyclonal
PBS with 0.09% sodium azide
P60981
ACTDP, ADF, bA462D18.2, actin-depolymerizing factor, bA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)), destrin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11034
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DSTN
Liquid
DSTN
KLH conjugated synthetic peptide between 70-98 amino acids from the central region of human DSTN
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit