Learn More
anti-Dtk, APC, Clone: 109646, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB759A
Catalog No. FAB759A
Add to cart
This item is not returnable. View return policy
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
Dtk/TYRO3
109646
Flow Cytometry 10 uL/10^6 cells
P55144
Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Rat
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse Dtk extracellular domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Dtk, recombinant mouse (rm) Mer, or rmAxl is observed.
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
Tyro3
Mouse myeloma cell line NS0-derived recombinant mouse Dtk Ala31-Ser418 Accession # P55144
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7301
Mouse