Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Dtk/TYRO3, Alexa Fluor 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128635AF405

Catalog No. NBP128635W


Add to cart

Description

Description

Dtk/TYRO3 Polyclonal antibody specifically detects Dtk/TYRO3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Dtk/TYRO3
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
TYRO3
Synthetic peptide made to an internal portion of the human TYRO3 protein (within residues 100-250). [Swiss-Prot# Q06418]
96 kDa
0.1 ml
Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases
Primary
7301
Human, Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Q06418
Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human and mouse. Immunogen sequence has 94% identity to rat.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only