Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dtk/TYRO3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP128635SS
Description
Dtk/TYRO3 Polyclonal specifically detects Dtk/TYRO3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).Specifications
Dtk/TYRO3 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY | |
Rabbit | |
96 kDa | |
0.025 mL | |
Neuroscience, Neurotransmission, Tyrosine Kinases | |
7301 | |
Human, Mouse | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
1.19 mg/mL | |
Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 27034374), Immunocytochemistry/Immunofluorescence 1:200, Flow (Intracellular) | |
Q06418 | |
TYRO3 | |
Synthetic peptide made to an internal portion of the human TYRO3 protein (within residues 100-250). [Swiss-Prot# Q06418] | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Immunogen sequence has 94% identity to rat. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction