Learn More
DTNB Mouse anti-Human, Clone: 1D3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518771
Catalog No. PIMA518771
Add to cart
Description
Description
Peptide Sequence: MIEESGNKRK TMAEKRQLFI EMRAQNFDVI RLSTYRTACK LRFVQKRCNL HLVDIWNMIE AFRDNGLNTL DHTTEISVSR LETVISSIY
This gene encodes dystrobrevin beta, a component of the dystrophin-associated protein complex (DPC). The DPC consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and dystrobrevin alpha and beta. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Dystrobrevin beta is thought to interact with syntrophin and the DP71 short form of dystrophin. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Specifications
DTNB
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
O60941
DTN-B, beta-dystrobrevin, dystrobrevin beta
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1838
ELISA
1D3
Unconjugated
DTNB
Liquid
DTNB
DTNB (NP_899205, 1 a.a. ∽ 89 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human