Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dystrobrevin, beta (B01), Mouse anti-Human, Rat, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human DTNB protein.

Manufacturer:  Abnova Corporation H00001838B01

Catalog No. 89-005-266


Add to cart

Description

Description

This gene encodes dystrobrevin beta, a component of the dystrophin-associated protein complex (DPC). The DPC consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and dystrobrevin alpha and beta. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Dystrobrevin beta is thought to interact with syntrophin and the DP71 short form of dystrophin. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Sequence: MIEESGNKRKTMAEKRQLFIEMRAQNFDVIRLSTYRTACKLRFVQKRCNLHLVDIWNMIEAFRDNGLNTLDHTTEISVSRLETVISSIYYQLNKRLHSTHQISVEQSISLLLNFMTAAYDSEGRGKLTVFSVKAMLATMCGGKMLDKLRYVFSQMSDSNGLMIFSKFDQFLKEVLKLPTAVFEGPSFGYTEHSVRTCFPQQRKIMLNMFLDTMMADPPPQCLVWLPLMHRLAHVENVFHPVECSYCRCESMMGFRYRCQQCHNYQLCQNCFWRGHAGGPHSNQHQMKEHSSWKSPAKKLSHAISKSLGCVPTREPPHPVFPEQPEKPLDLAHIVPPRPLTNMNDTMVSHMSSGVPTPTKRLQYSQDIPSHLADEHALIASYVARLQHCARVLDSPSRLDEEHRLIARYAARLAAEAGNVTRPPTDLSFNFDANKQQRQLIAELENKNREILQEIQRLRLEHEQASQPTPEKAQQNPTLLAELRLLRQRKDELEQRMSALQESRRELMVQLEELMKLLKEEEQKQAAQATGSPHTSPTHGGGRPMPMPVRSTSAGSTPTHCPQDSLSGVGGDVQEAFAQGTRRNLRNDLLVAADSITNTMSSLVKELHSG
Specifications

Specifications

dystrobrevin, beta
Unconjugated
No additive
BC049366.1
MGC17163|MGC57126
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1838
Western Blot
Mouse polyclonal antibody raised against a full-length human DTNB protein.
DTNB
Serum
DTNB
DTNB (AAH49366.1, 1 a.a. ~ 609 a.a) full-length human protein.
RUO
Y
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit