Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DTNBP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512283

Catalog No. PIPA512283

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Dtnp1 may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations in this protein are associated with Hermansky-Pudlak syndrome type 7. This protein may also be associated with schizophrenia.
Specifications

Specifications

Dtnbp1
Polyclonal
PBS with 0.09% sodium azide
Q96EV8, Q91WZ8
Dysbindin; Dystrobrevin-binding protein 1; Hermansky-Pudlak syndrome 7 protein homolog; Hps7-like protein; Dtnbp1; Sdy
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84062, 94245
Immunohistochemistry, Western Blot
Unconjugated
DTNBP1
Liquid
DTNBP1
KLH conjugated synthetic peptide between 296-325 amino acids from the C-terminal region of human Dtnbp1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit