Learn More
Seracare Life Sciences Inc ANTI-DUCK IGG (H+L) ANTIBODY
Supplier:  Seracare Life Sciences Inc 52100206
Encompass_Preferred
Affinity purified polyclonal antibody to Duck IgG, both heavy and light chains (whole IgG) made in Goat. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-668-47
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.