Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP13 Isoform 3 Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141643

Catalog No. 01-675-256


Add to cart

Description

Description

Suggested positive control: human skeletal muscle lysate.

DUSP13, a novel DSP, is a member of highly conserved Protein-tyrosine phosphatase super family. Members of this family cooperate with protein kinases and play a vital role in cell cycle regulation. DUSP13 consists of a conserved catalytic domain and is expressed specifically in the testis and skeletal muscle. Based on mRNA localization and expression studies during development, DUSP13 is suggested to be expressed as 3 isoforms due to alternative splicing. Reports suggest DUSP13 might be involved in the regulation of meiosis and differentiation of testicular germ cells during spermatogenesis.
Specifications

Specifications

DUSP13 Isoform 3
Polyclonal
PBS with 0.2% gelatin and 0.05% sodium azide
Q6B8I1
RP11-487I5.6, BEDP, DUSP13A, DUSP13B, FLJ32450, MDSP, SKRP4, TMDP
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
51207
Western Blot
Unconjugated
DUSP13
Liquid
DUSP13
Synthetic peptide corresponding to residues 50-100 of human DUSP13.
Protein G
RUO
Antibody
Polyclonal
Human, Non-human Primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit