Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DUSP19, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132399

Catalog No. NBP132399


Add to cart

Description

Description

DUSP19 Polyclonal antibody specifically detects DUSP19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DUSP19
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
DUSP19
Recombinant protein encompassing a sequence within the center region of human DUSP19. The exact sequence is proprietary.
24 kDa
100 ul
Protein Phosphatase
Primary
142679
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
dual specificity phosphatase 19, Dual specificity phosphatase TS-DSP1, dual specificity protein phosphatase 19, DUSP17SAPK pathway-regulating phosphatase 1, EC 3.1.3.16, EC 3.1.3.48, LMWDSP3, LMW-DSP3, Low molecular weight dual specificity phosphatase 3, Protein phosphatase SKRP1, SKRP1MGC138210, Stress-activated protein kinase pathway-regulating phosphatase 1, TS-DSP1
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only