Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKP3, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA128321

Catalog No. PIPA128321

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

DUSP6a inactivates MAP kinases. It has a specificity for the ERK family, and is implicated in muscle and neuronal differentiation.

PA1-28321 detects DUSP6a from human, mouse, rat samples.PA1-28321 has been successfully used in Western blot applications. Detects a band of approximately 60kDa in an E-coli fusion protein sample (predicted molecular weight: 42kDa).The PA1-28321 immunogen is synthetic peptide, corresponding to amino acids 135-280 of Human DUSP6a.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DUSP6a
Polyclonal
Unconjugated
Q16828, Q9DBB1, Q64346
DUSP6
Synthetic peptide, corresponding to amino acids 135-280 of Human DUSP6a.
50 μg
Primary
Human, Mouse, Rat
Antibody
IgY
Western Blot
1mg/mL
DUSP6
MKP3, PYST1
Chicken
Antigen affinity chromatography
RUO
1848
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit