Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

DVL3, Mouse, MaxPab™ Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human DVL3 protein.

Manufacturer:  Abnova Corporation H00001857B01

Catalog No. 89-002-676



Description

Description

This gene is a member of a multi-gene family which shares strong similarity with the Drosophila dishevelled gene, dsh. The Drosophila dishevelled gene encodes a cytoplasmic phosphoprotein that regulates cell proliferation. [provided by RefSeq

Sequence: MGETKIIYHLDGQETPYLVKLPLPAERVTLADFKGVLQRPSYKFFFKSMDDDFGVVKEEISDDNAKLPCFNGRVVSWLVSAEGSHPDPAPFCADNPSELPPPMERTGGIGDSRPPSFHPHAGGGSQENLDNDTETDSLVSAQRERPRRRDGPEHATRLNGTAKGERRREPGGYDSSSTLMSSELETTSFFDSDEDDSTSRFSSSTEQSSASRLMRRHKRRRRKQKVSRIERSSSFSSITDSTMSLNIITVTLNMEKYNFLGISIVGQSNERGDGGIYIGSIMKGGAVAADGRIEPGDMLLQVNEINFENMSNDDAVRVLREIVHKPGPITLTVAKCWDPSPRGCFTLPRSEPIRPIDPAAWVSHTAAMTGTFPAYGMSPSLSTITSTSSSITSSIPDTERLDDFHLSIHSDMAAIVKAMASPESGLEVRDRMLLKITIPNAFIGSDVVDWLYHNVEGFTDRREARKYASNLLKAGFIRHTVNKITFSEQCYYIFGDLCGNMANLSLHDHDGSSGASDQDTLAPLPHPGAAPWPMAFPYQYPPPPHPYNPHPGFPELGYSYGGGSASSQHSEGSRSSGSNRSGSDRRKEKDPKAGDSKSGGSGSESDHTTRSSLRGPRERAPSERSGPAASEHSHRSHHSLASSLRSHHTHPSYGPPGVPPLYGPPMLMMPPPPAAMGPPGAPPGRDLASVPPELTASRQSFRMAMGNPSEFFVDVM
Specifications

Specifications

DVL3
Unconjugated
No additive
BC032459
KIAA0208
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human DVL3 protein.
DVL3
Liquid
DVL3
DVL3 (AAH32459, 1 a.a. ~ 716 a.a) full-length human protein.
RUO
Primary
1857
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit