Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
UbpbioSupplier Diversity Partner ANTI-DYKDDDDK, MOUSE 100UG DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner Y1101
Antibodies – Anti-DYKDDDDK (UBPBio catalog# Y1091) – Mouse monoclonal antibody (clone GF.4R, IgG1). Also called FLAG antibody. Used for immunoblotting at 1:1,000-3,000 dilution for detection of 1-5 ng FLAG-tagged protein. 1:500-2,000 dilution for immunostaining, 1-3 ug FLAG antibody/reaction for immunoprecipitation.
Catalog No. 50-112-6925