Learn More
DYM Mouse anti-Human, Clone: 1C2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522064
Catalog No. PIMA522064
Add to cart
Description
Description
Peptide Sequence: QTSDQATLLL YTLLHQNSNI RTYMLARTDM ENLVLPILEI LYHVEERNSH HVYMALIILL ILTEDDGFNR SIHEVILKNI TWYSERVL
This gene encodes a protein which is necessary for normal skeletal development and brain function. Mutations in this gene are associated with two types of recessive osteochondrodysplasia, Dyggve-Melchior-Clausen (DMC) dysplasia and Smith-McCort (SMC) dysplasia, which involve both skeletal defects and mental retardation.
Specifications
Specifications
DYM
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q7RTS9
DYM
DYM (NP_060123, 343 a.a. ∽ 430 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human
ELISA
1C2
Unconjugated
DYM
Liquid
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
54808