Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynactin p50 Mouse, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611003

 View more versions of this product

Catalog No. BDB611003


Add to cart

Description

Description

Dyneins are ubiquitous, multimeric proteins that are responsible for minus end-directed microtubule-based organelle transport. They are divided into two classes, cytosolic and axonemal. The activity of cytosolic dynein is regulated by the multiprotein dynactin complex with which it co-purifies. Dynactin is composed of at least nine polypeptides with p50 being the second most abundant subunit. The complex consists of an F-actin-like core filament of actin-related protein Arp1, a heterodimer that acts as a cap at one end, and a 62kDa subunit at the other end. p150[glued] is the largest subunit of the complex. The precise function of dynactin remains to be determined, but it may be recruited to prometaphase kinetochores to assist in alignment and spindle organization during mitosis. Additionally, dynactin localizes at the tail of cytosolic dynein and activates dynein-mediated vesicle movement. Therefore, dynactin may function to anchor or target dynein to the kinetochore during mitosis.
Specifications

Specifications

Dynactin p50
25
Unconjugated
Affinity Purified
Human Dynactin p50 aa. 55-196
150μg
Cell Biology
Primary
Drosophila, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.