Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamin Mouse, Unlabeled, Clone: 41, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610245

 View more versions of this product

Catalog No. BDB610245


Add to cart

Description

Description

Dynamin is a nerve terminal phosphoprotein with intrinsic GTPase activity which plays an important role in endocytosis. This GTPase activity is stimulated in vitro by microtubules, SH3 domain-containing proteins, phospholipids, and Protein Kinase C (PKC)-mediated phosphorylation. There are at least two distinct dynamin genes in mammals, which show 79% identity, encoding proteins Dynamin I and Dynamin II. Dynamin I is expressed almost exclusively in the central nervous system while Dynamin II expression is ubiquitous. Depolarization stimulates synaptic vesicle recycling and Dynamin I is subsequently dephosphorylated. This rapid dephosphorylation is mediated by calcineurin which acts as a switch for depolarization-initiated synaptic vesicle endocytosis. The C-terminus of dynamin contains two clusters of proline-rich SH3 domain binding proline motifs which interact with known SH3 domain proteins during tyrosine kinase receptor activation. It is reported that clone 41 detects both dynamin I and dynamin II.

Specifications

Specifications

Dynamin
41
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.