Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamin II Mouse, Unlabeled, Clone: 27, BD

Mouse Monoclonal Antibody

$226.00

Specifications

Antigen Dynamin II
Isotype IgG2a
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610263
View Documents Promotion Details
BD Biosciences
610263
50μg
Each for $226.00
Add to cart
 
BDB610264
View Documents Promotion Details
BD Biosciences
610264
150μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Dynamin is a membrane-associated GTPase that binds to GTP, microtubules, and phospholipids. Northern blot analysis has shown dynamin to be present in most tissues but at an amount about 20-fold lower than that in brain. There are at least two distinct dynamin genes in mammals. Transcripts of both dynamin genes are alternatively spliced at two or more sites. The first site is identical in bothdynamins, whereas, the second site differs. The two gene products are known as Dynamin I and Dynamin II and show 79% identity. Dynamin I is expressed almost exclusively in the central nervous system while Dynamin II expression is ubiquitous. The two proteins are highly homologous in the N-terminal region, while the C-terminal domain shows significant divergence. The GTPase activity of Dynamin I is stimulated several fold by binding to microtubules, phospholipids, and membranous vesicles. Dynamin I is a good substrate of PKC in vitro as well as in vivo in resting nerve terminals. In vitro, phosphorylation occurs at the C-terminus of Dynamin I and this enhances the GTPase activity more than 10-fold. However, Dynamin II is not a substrate of PKC and its activity does not appear to be affected by phosphorylation. These data suggest that the function of the common N-terminal domain between Dynamin I and II may be differentially specified by distinct C-terminal domains.

Specifications

Specifications

Dynamin II
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG2a
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Immunofluorescence, Western Blot
RUO
27
250μg/mL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit