Learn More
α-Dystroglycan Mouse anti-Human, Rat, Mouse, Rabbit, Canine, Guinea Pig, Clone: IIH6C4, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05593
Catalog No. 05-593-MI
Add to cart
Description
Description
This Anti-α-Dystroglycan Antibody, clone IIH6C4 is validated for use in immunohistochemistry, FUNC, western blotting for the detection of α-Dystroglycan.
Specifications
Specifications
α-Dystroglycan
IIH6C4
Mouse ascites, in PBS containing 0.05% sodium azide and 30% glycerol. Liquid at −20deg;C.
Ascites
Murine
IgM
200μL
Primary
Canine, Guinea Pig, Human, Murine, Rabbit, Rat
Functional Assay, Immunohistochemistry, Western Blot
Unconjugated
NM_004393
DAG1, A3a, DAG, beta-dystroglycan, alpha-dystroglycan, AGRNR, 156DAG
Rabbit skeletal muscle membrane preparation. Clone IIH6C4.
Unpurified
−20°C for up to 12 months.
Monoclonal