Learn More
Dystrophin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA121011
Catalog No. PA121011
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Dystrophin is a member of the spectrin/alpha-actinin family of actin-binding, triple helix rod-containing proteins. It is absent or greatly reduced in individuals with the X-linked recessive Duchenne's muscular dystrophy disorder, as well as in mice with the mdx (murine muscular dystrophy) mutation.
PA1-21011 detects Dystrophin in Human, Mouse and Rat samples.PA1-21011 has been successfully used in Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) immunocytochemistry, immunofluorescence, and Western Blot procedures. Staining of formalin-fixed tissues requires boiling tissue sections in 1 mM EDTA, pH 8.0 for 10 minutes followed by cooling at room temperature for 20 minutes. Positive control of skeletal muscle suggested. Molecular weight of 427kDa is expected. PA1-21011 immunogen corresponds to a synthetic peptide corresponding to C-terminal of human Dystrophin.
Specifications
Specifications
Dystrophin
Polyclonal
Unconjugated
DMD
Liquid
DMD
A synthetic peptide corresponding to C-terminal of human Dystrophin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
0.2mg/mL
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
P11532, P11531, P11530
GS1-19O24.1, BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272
Rabbit
IgG
500μL
4° C, do not freeze, do not freeze
Primary
13405, 1756, 24907