Learn More
E-Cadherin Mouse, FITC, Clone: 36/E-CADHERIN, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612131
Description
E-Cadherin is a 120kDa transmembrane glycoprotein that is localized in the adherens junctions of epithelial cells. There, it interacts with the cytoskeleton through the associated cytoplasmic catenin proteins. In addition to being a calcium-dependent adhesion molecule, E-Cadherin is also a critical regulator of epithelial junction formation. Its association with catenins is necessary for cell-cell adhesion. These E-cadherin/catenin complexes associate with corical actin bundles at both the zonula adherens and the lateral adhesion plaques. Tyrosine phosphorylation can disrupt these complexes, leading to changes in cell adhesion properties. E-Cadherin expression is often down-regulated in highly invasive, poorly differentiated carcinomas. Increased expression of E-Cadherin in these cells reduces invasiveness. Thus, loss of expression or function of E-Cadherin appears to be an important step in tumorigenic progression.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Specifications
E-Cadherin | |
Monoclonal | |
250μg/mL | |
CD324; CDH1; CADH1; Cadherin-1; ECAD; CDHE; Arc-1; LCAM; UVO; Uvomorulin | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Immunofluorescence | |
36/E-CADHERIN | |
FITC | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human E-Cadherin aa. 735-883 | |
150 μg | |
Cell Biology | |
Canine, Human, Mouse, Rat | |
IgG2a κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.