Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-E-Cadherin, Fluorescein, Clone: 114420, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Specifications
Antigen | E-Cadherin |
---|---|
Clone | 114420 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB7481F025
|
R&D Systems
FAB7481F025 |
25 Tests | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
FAB7481F100
|
R&D Systems
FAB7481F100 |
100 Tests | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Mouse samples. It is validated for Flow Cytometry.Specifications
E-Cadherin | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin | |
CDH1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse E-Cadherin in direct ELISAs and Western blots. |
114420 | |
Flow Cytometry | |
Fluorescein | |
Rat | |
Mouse | |
P09803 | |
999 | |
Mouse myeloma cell line NS0-derived recombinant mouse E-Cadherin Asp157-Val709 (predicted) Accession # P09803 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title