Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

E. coli serotype 987P Pili Sheep anti-Bacteria, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA185409

Catalog No. PA185409



Description

Description

Sheep anti Escherichia coli 987P pilus antibody recognizes the E. coli 987P pilus, a structure that mediates attachment and subsequent colonization of host surfaces.

Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
Specifications

Specifications

E. coli serotype 987P Pili
Polyclonal
Unconjugated
Liquid
Sheep
IgG
1 mL
4°C or -20°C if preferred
Primary
Bacteria
ELISA
5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Escherichia coli
E. coli strain CVL Moon 987P O:9K103 was used.
Affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit