Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

E. coli serotype O/K Goat anti-Bacteria, FITC, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA173033

Catalog No. PA173033



Description

Description

The PA1-73033 antibody reacts with E. coli. PA1-73033 has been successfully used in immunofluorescence applications. The PA1-73033 antibody was raised against E. coli serotypes O157:H7, O20, O125, O55, O111 & K12.

Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
Specifications

Specifications

E. coli serotype O/K
Polyclonal
FITC
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Bacteria
Immunofluorescence
4-5 mg/mL
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Escherichia coli
E. coli serotypes O157:H7, O20, O125, O55, O111 & K12.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit