Learn More
E. coli serotype K99 Pili Mouse anti-Bacteria, Clone: K9912-444.5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110798
Catalog No. 01-671-827
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10798 detects E. coli K99 pili. MA1-10798 has been successfully used in ELISA applications. The MA1-10798 immunogen is purified E. coli K99 pili.
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
Specifications
Specifications
E. coli serotype K99 Pili
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Escherichia coli
Purified E. coli K99 pili.
Purified
RUO
Antibody
Monoclonal
ELISA
K9912-444.5
Unconjugated
Liquid
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Bacteria