Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. coli serotype O/K, Biotin, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA128963

Catalog No. PIPA128963

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.

PA1-28963 detects E. coli O+K from bacteria samples.PA1-28963 has been successfully used in immunofluorescence applications.The PA1-28963 immunogen is: Tissue / cell preparation (Mixture of E. coli serotypes).
Specifications

Specifications

E. coli serotypes O + K
Polyclonal
Biotin
Purified
Goat
IgG
100μL
Store at -20°C
Primary
Bacteria
Immunocytochemistry, Immunofluorescence
4mg/mL
PBS with 0.1% sodium azide; pH 7.2
Escherichia coli
Tissue/cell preparation (Mixture of E. coli serotypes).
IgG fraction
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit