Learn More
E. coli serotype O/K, HRP, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA127375
Catalog No. PA127375
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
PA1-27375 detects E. coli of all O and K antigenic serotypes from bacteria samples.PA1-27375 has been successfully used in ELISA procedures.The PA1-27375 immunogen is a mixture of E. coli serotypes.
Specifications
Specifications
E. coli serotypes O + K
Polyclonal
HRP
Purified
Goat
IgG
100μL
Store at -20°C
Primary
Bacteria
ELISA, Immunocytochemistry, Immunofluorescence
1mg/mL
PBS with 10mg/mL BSA and 0.002% thimerosal
Escherichia coli
Mixture of E. coli serotypes.
IgG fraction
RUO
Antibody
Polyclonal