Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-E COLI O111 SPECIES AB  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 53300070
Affinity purified polyclonal antibody to E. coli O111:H8 made in goat labeled with DyLight 488. Isolated from a serum pool of goats immunized with heat-killed whole cells of E. coli serotype O111. The antibody is highly specific for E. coli O111:H8. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay
Catalog No. 50-670-97