Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-E COLI O157 H7 BIOTINANT  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 53600032
Affinity purified antibody isolated from a pool of serum from goats immunized with Eschericia coli O157:H7 was labeled with a biotin analog having a long carbon spacer arm. This minimizes steric hindrance and maximizes binding.
Catalog No. 50-672-95