Learn More
E. coli serotype O/K Rabbit anti-Bacteria, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA125636
Catalog No. PA125636
Add to cart
Description
Description
PA1-25636 detects E. coli of all "O" and "K" antigenic serotypes from bacteria samples. PA1-25636 has been successfully used in immunofluorescence, immunocytochemistry, and ELISA procedures. The PA1-25636 immunogen is a mixture of all antigenic serotypes. Store at 4°C short term. For long term storage, aliquot and store at -20°C, avoiding freeze/thaw cycles.
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
Specifications
Specifications
E. coli serotype O/K
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
500 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Bacteria
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
4 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Escherichia coli
Mixtures of all antigenic serotypes.
IgG fraction
RUO
Antibody
Polyclonal