Learn More
E. coli serotype O/K Goat anti-Bacteria, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA127226
Catalog No. PA127226
Add to cart
Description
Description
PA1-27226 detects E. coli from bacteria samples. It is reactive to all E.Coli species in the immunogen, plus 0157-H7 and K12 tested by immunoblot, reacts with more then 50 proteins by Western Blot PA1-27226 has been successfully used in ICC, Western blot and ELISA procedures. The PA1-27226 immunogen is heat-killed sonicate blend of 6 different strains of E. coli: TOP 10F, XS127/P3, HB101, JM109, CJ236/P3, and ATCC 37197. Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20° C or below, avoiding multiple freeze-thaw cycles.
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
Specifications
Specifications
E. coli serotype O/K
Polyclonal
Unconjugated
Liquid
Goat
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Bacteria
ELISA, Immunocytochemistry, Immunofluorescence
4 mg/mL
0.01M PBS with 0.1% sodium azide; pH 7.2
Escherichia coli
Heat killed sonicate blend of 6 different strains of E. coli: TOP 10F, XS127/P3, HB101, JM109, CJ236/P3, and ATCC 37197.
IgG fraction
RUO
Antibody
Polyclonal