Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. coli serotype O/K Rabbit anti-Bacteria, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA17213

Catalog No. PA17213


Add to cart

Description

Description

PA1-7213 detects all "O" and "K" antigenic serotypes of Escherichia coli. This antibody is unabsorbed and will cross react with related enterobacteriaceae. PA1-7213 is suggested for use in ELISA procedures. The PA1-7213 immunogen is mixtures of all antigenic serotypes. Recommended short term (6 months) storage is liquid at 2-8 °C. For longer term storage, aliquot and freeze.

Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
Specifications

Specifications

E. coli serotype O/K
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Bacteria
ELISA
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Escherichia coli
Mixtures of all antigenic serotypes.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit