Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
E-selectin Rabbit anti-Human, Non-human primate, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA140301
Catalog No. 01-676-420
Description
Description
PA1-40301 detects E-Selectin from human, chimpanzee, rhesus monkey, cynomolgous monkey and baboon samples. PA1-40301 reacts with the Lectin Domain of E-selectin, CD62-e, formerly designated Endothelial Leucocyte Adhesion Molecule-1 (ELAM-1). PA1-40301 reacts with human endothelial cells activated with TNF, IL-1 or endotoxin. PA1-40301 was found to react also with cells transfected with the ELAM-1 gene. The antibody inhibits the adhesion of granulocytes both neutrophilic and eosinophilic. PA1-40301 has been successfully used in ELISA, immunohistochemistry (frozen) and Western blot applications. It is recommended that users test the reagent and determin their own optimal dilutions: the typical starting working dilution is 1:10. The PA1-40301 imunogen is human E-Selectin.
Lectin Domain of E-selectin, CD62-e, formerly designated Endothelial Leucocyte Adhesion Molecule-1 (ELAM-1).
Specifications
Specifications
E-selectin
Polyclonal
Biotin
SELE
Liquid
SELE
Human E-Selectin.
Protein A
RUO
Antibody
Polyclonal
Human, Non-human Primate
ELISA, Immunohistochemistry (Frozen), Western Blot
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
P16581
RP1-117P20.2, CD62E, ELAM, ELAM1, ESEL, LECAM2
Rabbit
IgG
50 μg
4° C
Primary
6401