Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E Tag Rabbit anti-Tag, HRP, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA121895

Catalog No. PA121895


Add to cart

Description

Description

PA1-21895 detects E tag. PA1-21895 has been successfully used in ELISA and Western Blot procedures. The PA1-21895 immunogen is a synthetic peptide corresponding to residues GAPVPYPDPLEPR, conjugated with KLH.

Specifications

Specifications

E Tag
Polyclonal
HRP
Liquid
Rabbit
IgG
100 μg
4° C
Primary
Tag
ELISA, Western Blot
1 mg/mL
HEPES buffered saline with 0.1M NaCl, 0.2% BSA and 0.01% thimerosal; pH 7.0
E epitope tag; GAPVPYPDPLEPR; GAPVPYPDPLEPR tag
Synthetic peptide corresponding to residues GAPVPYPDPLEPR, conjugated with KLH.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit