Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-E2F-1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131394

Catalog No. NBP131394


Add to cart

Description

Description

E2F-1 Polyclonal antibody specifically detects E2F-1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

E2F-1
1.0mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Q01094
E2F transcription factor 1, E2F-1, PBR3, PRB-binding protein E2F-1, RBAP1, RBAP-1, RBBP3, RBBP-3, RBP3, Retinoblastoma-associated protein 1, Retinoblastoma-binding protein 3, transcription factor E2F1
Rabbit
IgG
100 ul
Apoptosis, Cancer, Cell Cycle and Replication, Transcription Factors and Regulators
Primary
1869
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
E2F1
Recombinant protein encompassing a sequence within the center region of human E2F1. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only