Learn More
E2F-3 Mouse anti-Human, Mouse, Clone: PG37, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05551
Catalog No. 05-551-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect E2F-3 with Anti-E2F-3 Antibody, clone PG37 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation, western blot, immunocytochemistry.
Specifications
Specifications
E2F-3
PG37
PBS, pH 7.4
Purified
Murine
IgG2a
100μg
Primary
Human, Murine
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_001949
E2F3, KIAA0075, E2F-3, MGC104598, DKFZp686C18211
Recombinant full-length human E2F-3 protein
Protein A purfied
2-8°C for one year
Monoclonal