Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F-4 Mouse anti-Human, Mouse, Rat, Clone: GG22-2A6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05312
Description
Detect E2F-4 with Anti-E2F-4 Antibody, clone GG22-2A6 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation and western blotting.Specifications
E2F-4 | |
Monoclonal | |
Unconjugated | |
NM_001950 | |
Mouse | |
Protein G chromatography | |
Primary | |
−20°C for one year | |
IgG2b |
Immunoprecipitation, Western Blot | |
GG22-2A6 | |
200μg of protein G purified mouse IgG2b in 200μL of 0.07MTris-glycine, pH 7.4, 0.135MNaCl, 0.035% sodium azide and 30% glycerol. | |
E2F4, E2F-4 | |
GST fusion to human E2F-4 | |
200 μg | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only