Learn More
E2F4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA129136
Catalog No. PIPA129136
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-29137 detects E2F5 from mouse, human, and rat samples. PA1-29137 has been successfully used in Western blot applications. Suggested positive controls for this product are MCF-7, NIH/3T3 cells The PA1-29137 immunogen is synthetic peptide correspondin
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein binds to all three of the tumor suppressor proteins pRB, p107 and p130, but with higher affinity to the last two. It plays an important role in the suppression of proliferation-associated genes, and its gene mutation and increased expression may be associated with human cancer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E2F4
Polyclonal
Unconjugated
Q16254, Q8R0K9
E2F4
Synthetic peptide from the C terminus of human E2F4 protein.
500 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
1 mg/mL
E2F4
E2F-4
Rabbit
Protein A
RUO
100360427, 104394, 1874
4° C, do not freeze
Liquid
Recommendations
Recommendations