Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F6 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA519143
Description
This antibody is predicted to react with bovine and canine based on sequence homology.
E2F6 Polyclonal antibody specifically detects E2F6 in Human samples. It is validated for Western Blot.Specifications
E2F6 | |
Polyclonal | |
Unconjugated | |
E2F6 | |
AI462434; E2F transcription factor 6; E2f6; E2F-6; E2F6b; E2F-binding site-modulating activity protein; EMA; MGC111545; Transcription factor E2F6; transcriptional repressor E2F6 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
1876 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O75461 | |
E2F6 | |
Synthetic peptide sequence (NPQQSEELLEVSN) corresponding to the C-terminus amino acids of E2F6 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction