Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F8, Goat, Polyclonal Antibody, Abnova
Goat polyclonal antibody raised aganist synthetic peptide of E2F8.
Manufacturer: Abnova Corporation PAB15624
Description
E2F transcription factors, such as E2F8, are essential for orchestrating expression of genes required for cell cycle progression and proliferation (Christensen et al., 2005 [PubMed 16179649]).[supplied by OMIM
Sequence: C-KSIESDRRKINSAPSSpecifications
E2F8 | |
0.5 mg/mL | |
Goat polyclonal antibody raised aganist synthetic peptide of E2F8. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_078956.2 | |
FLJ23311 | |
Goat | |
Antigen affinity purification | |
RUO | |
Primary | |
79733 |
ELISA | |
Unconjugated | |
ELISA (1:32000) The optimal working dilution should be determined by the end user. | |
E2F8 | |
Liquid | |
E2F8 | |
A synthetic peptide corresponding to amino acids at internal region of human E2F8. | |
100 ug | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only