Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E4F transcription factor 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human E4F1 protein.

Manufacturer:  Abnova Corporation H00001877B01

Catalog No. 89-005-273


Add to cart

Description

Description

The zinc finger protein encoded by this gene is one of several cellular transcription factors whose DNA-binding activities are regulated through the action of adenovirus E1A. A 50kDa amino-terminal product is generated from the full-length protein through proteolytic cleavage. The protein is differentially regulated by E1A-induced phosphorylation. The full-length gene product represses transcription from the E4 promoter in the absence of E1A, while the 50kDa form acts as a transcriptional activator in its presence. [provided by RefSeq

Sequence: MEGAMAVRVTAAHTAEAQAEAGREAGEGAVAAVAAALAPSGFLGLPAPFSEEDEDDVHRCGRCQAEFTALEDFVQHKIQKACQRAPPEALPATPATTALLGQEVVPAAPGPEEPITVAHIVVEAASLAADISHASDLVGGGHIKEVIVAAEAELGDGEMAEAPGSPHQQGLGLAGEGEQAQVKLLVNKDGRYVCALCHKTFKTGSILKAHMVTHSSRKDHECKLCGASFRTKGSLIRHHRRHTDERPYKCSKCGKSFRESGALTRHLKSLTPCTEKIRFSVSKDVVVSKEDARAGSGAGAAGLGTATSSVTGEPIETSPVIHLVTDAKGTVIHEVHVQMQELSLGMKALAPEPPVSQELPCSSEGSRENLLHQAMQNSGIVLERAAGEEGALEPAPAAGSSPQPLAVAAPQLPVLEVQPLETQVASEASAVPRTHPCPQCSETFPTAATLEAHKRGHTGPRPFACAQCGKAFPKAYLLKKHQEVHVRERRFRCGDCGKLYKTIAHVRGHRRVHSDERPYPCPKCGKRYKTKNAQQVHFRTHLEEKPHVCQFCSRGFREKGSLVRHVRHHTGEKPFKCYKCGRGFAEHGTLNRHLRTKGGCLLEVEELLVSEDSPAAATTVLTEDPHTVLVEFSSVVADTQEYIIEATADDAETSEATEIIEGTQTEVDSHIMKVVQQIVHQASAGHQIIVQNVTMDEETALGPEAAAADTITIATPESLTEQVAMTLASAISEGTVLAARAGTSGTEQATVTMVSSEDIEILEHAGELVIASPEGQLEVQTVIV
Specifications

Specifications

E4F transcription factor 1
Unconjugated
No additive
BC080524.1
E4F|MGC99614
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1877
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human E4F1 protein.
E4F1
Serum
E4F1
E4F1 (AAH80524.1, 1 a.a. ~ 784 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit