Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14393

Catalog No. PA14393


Add to cart

Description

Description

PA1-4393 detects EAPII from human samples. PA1-4393 has been successfully used in Western blot procedures. PA1-4393 immunizing peptide corresponds to amino acid residues 11-28 from human EAPII protein.

Ets proteins constitute a family of conserved sequence-specific DNA-binding proteins and function as transcription factors with roles in differentiation, lymphoid cell development, invasiveness and angiogenesis. A novel ETS1 interacting protein designated as ETS1-associated protein II (EAPII) interacts with ETS1 and other Ets proteins (ETS2 and FLI1) and is a novel ETS1 modulator.
Specifications

Specifications

EAP2
Polyclonal
Unconjugated
TDP2
Liquid
TDP2
Synthetic Peptide: R(11) E A A E E E G E P E V K K R R L L(28)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O95551
TRAF and TNF receptor-associated protein, ETS1-associated protein 2, ETS1-associated protein II, EAPII
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
51567
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit